cis-1,3-Cyclohexanedicarboxylic acid ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Warning

Synonymer: cis-Cyclohexane-1,3-dicarboxylic acid

C2197-25G C2197-5G
TCIAC2197-25GEA 4850 NOK
TCIAC2197-25G TCIAC2197-5G
cis-1,3-Cyclohexanedicarboxylic acid ≥98.0% (by GC, titration analysis)
cis-1,3-Cyclohexanedicarboxylic acid
Formel: C₈H₁₂O₄
MDL nummer: MFCD00134411
CAS-nummer: 2305-31-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=04d981be2a6500f8f1ac96e90613c14b1344c4b1&vid=04d981be2a6500f8f1ac96e90613c14b1344c4b1&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 163.0 to 167.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR