Cefazolin sodium salt ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI

C2242-25G C2242-5G
TCIAC2242-25GEA 2400 NOK
TCIAC2242-25G TCIAC2242-5G
Cefazolin sodium salt ≥98.0% (by HPLC, titration analysis)
Cefazolin sodium salt
Formel: C₁₄H₁₄N₈NaO₄S₃
Smeltepunkt: 190°C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00056883
CAS-nummer: 27164-46-1
EINECS: 248-278-4
Merck Indeks: 14,01917

Bestill nå

Spesifikasjon Testresultater

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D -15.0 to -24.0 deg (C=10, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR