N-Carbobenzoxy-L-valine succinimidyl ester ≥98.0% (by HPLC)

Leverandør: TCI

Synonymer: N-Benzyloxycarbonyl-L-valine N-succinimidyl ester, (S)-2,5-Dioxopyrrolidin-1-yl 2-(((benzyloxy)carbonyl)amino)-3-methylbutanoate, N-Cbz-L-valine Succinimidyl Ester, Z-Val-OSu

C2334-5G
TCIAC2334-5GEA 610 NOK
TCIAC2334-5G
N-Carbobenzoxy-L-valine succinimidyl ester ≥98.0% (by HPLC)
N-Carbobenzoxy-L-valine succinimidyl ester
Formel: C₁₇H₂₀N₂O₆
MW: 348.36 g/mol
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00053547
CAS-nummer: 3496-11-5
EINECS: 222-493-3

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 114.0 to 118.0 deg-C
Specific rotation [a]20/D -23.0 to -26.0 deg (C=1, Dioxane)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR