Citalopram hydrobromide ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI

C2370-1G C2370-5G
TCIAC2370-1GEA 1100 NOK
TCIAC2370-1G TCIAC2370-5G
Citalopram hydrobromide ≥98.0% (by HPLC, titration analysis)
Citalopram hydrobromide
Formel: C₂₀H₂₁FN₂O
MDL nummer: MFCD02101306
CAS-nummer: 59729-32-7
EINECS: 261-890-6
Merck Indeks: 14,02318

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR