Kobalt(II)klorid heksahydrat ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Danger

Synonymer: Cobalt Chloride, Hexahydrate, Cobalt dichloride hexahydrate, Cobalt Chloride, 6-Hydrate, Cobaltous chloride hexahydrate

C2388-250G
TCIAC2388-250GEA 745 NOK
TCIAC2388-250G
Kobalt(II)klorid heksahydrat ≥98.0% (by titrimetric analysis)
Kobalt(II)klorid heksahydrat

CAS Number: 7791-13-1
MDL Number: MFCD00149652
Molecular Formula: Cl2Co
Molecular Weight: 129.83
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 87

Formel: CoCl₂·6H₂O
Kokepunkt: 110 °C (1013 hPa)
Smeltepunkt: 87 °C
Tetthet: 1,92 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00149652
CAS-nummer: 7791-13-1
EINECS: 231-589-4
FN: 3077
ADR: 9,III
Merck Indeks: 13,02462

Bestill nå

Spesifikasjon Testresultater

Purity/Analysis Method >98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR