(1R,3S)-3-(Carbobenzoxyamino)cyclohexanecarboxylic acid ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI

Synonymer: (1R,3S)-3-(Cbz-amino)cyclohexanecarboxylic Acid

C2400-100MG
TCIAC2400-100MGEA 3175 NOK
TCIAC2400-100MG
(1R,3S)-3-(Carbobenzoxyamino)cyclohexanecarboxylic acid ≥98.0% (by HPLC, titration analysis)
(1R,3S)-3-(Carbobenzoxyamino)cyclohexanecarboxylic acid
Formel: C₁₅H₁₉NO₄
MW: 277,32 g/mol
CAS-nummer: 1259278-10-8

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Optical purity (LC) min. 95.0 ee%
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR