Krystall fiolett for biochemical research

Leverandør: TCI
Danger

Synonymer: CI 42555

C2460-5G
TCIAC2460-5GEA 300 NOK
TCIAC2460-5G
Krystall fiolett for biochemical research
Krystall fiolett
Formel: C₂₅H₃₀ClN₃
Smeltepunkt: 211°C
MDL nummer: MFCD00011750
CAS-nummer: 548-62-9
EINECS: 208-953-6
FN: 3077
ADR: 9,III
Merck Indeks: 14,04395
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=faebb1841dbe0a5dc8f857d6869bfa297142c3ba&vid=6d00940fd8eef621129559c76c92cca74bf2d814&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Green to Dark green powder to crystal
Absorbance min. 0.300 (2mg/L, 588.0 to 592.0 nm) (calcd.on dried substance)
Cell staining test to pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR