5-Carboxyfluorescein, hydrate ≥97.0% (by HPLC, titration analysis)

Leverandør: TCI
Warning

Synonymer: Spiro(isobenzofuran-1(3H),9'-(9H)xanthene)-5-carboxylic acid, 3',6'-dihydroxy-3-oxo-, 5-FAM, 3',6'-Dihydroxy-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-5-carboxylic acid, 5-Carboxyfluorescein hydrate

C2477-100MG
TCIAC2477-100MGEA 1075 NOK
TCIAC2477-100MG
5-Carboxyfluorescein, hydrate ≥97.0% (by HPLC, titration analysis)
5-Carboxyfluorescein
Formel: C₂₁H₁₂O₇.xH₂O
MDL nummer: MFCD00036874
CAS-nummer: 76823-03-5

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 97.0 % (calcd.on anh.substance)
Water max. 10.0 %

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR