6-Carboxyfluorescein ≥97.0% (by HPLC)

Leverandør: TCI
Warning

Synonymer: 3',6'-Dihydroxy-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-6-carboxylic acid, 6-FAM

C2478-100MG
TCIAC2478-100MGEA 1200 NOK
TCIAC2478-100MG
6-Carboxyfluorescein ≥97.0% (by HPLC)
6-Carboxyfluorescein
Formel: C₂₁H₁₂O₇
MDL nummer: MFCD00036873
CAS-nummer: 3301-79-9

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 95.0 % (calcd.on anh.substance)
Water max. 10.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR