1,2,3,4-Cyclobutanetetracarboxylic acid ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Warning

C2502-1G
TCIAC2502-1GEA 2950 NOK
TCIAC2502-1G
1,2,3,4-Cyclobutanetetracarboxylic acid ≥98.0% (by GC, titration analysis)
1,2,3,4-Cyclobutanetetracarboxylic acid
Formel: C₈H₈O₈
MW: 232,14 g/mol
MDL nummer: MFCD00013270
CAS-nummer: 53159-92-5

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 % (total of isomer)
Purity (Neutralization titration) min. 98.0 %
Melting point 237.0 to 241.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR