3-Cyano-4-fluorophenylboronic acid (contains varying amounts of Anhydride)

Leverandør: TCI
Warning

Synonymer: 3-Cyano-4-fluorobenzeneboronic acid

C2676-1G
TCIAC2676-1GEA 1675 NOK
TCIAC2676-1G
3-Cyano-4-fluorophenylboronic acid (contains varying amounts of Anhydride)
3-Cyano-4-fluorophenylboronic acid
Formel: C₇H₅BFNO₂
MW: 164,93 g/mol
MDL nummer: MFCD03095130
CAS-nummer: 214210-21-6

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 113.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR