ɑ-Cyano-3-hydroxycinnamic acid ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Warning

Synonymer: 2-Cyano-3-(3-hydroxyphenyl)acrylic acid, α-Cyano-3-hydroxycinnamic acid, 2-Cyano-3-(3-hydroxyphenyl)acrylic Acid

C2677-5G
TCIAC2677-5GEA 2550 NOK
TCIAC2677-5G
ɑ-Cyano-3-hydroxycinnamic acid ≥98.0% (by HPLC, titration analysis)
alpha-Cyano-3-hydroxycinnamic acid
Formel: C₁₀H₇NO₃
MW: 189,17 g/mol
MDL nummer: MFCD00004203
CAS-nummer: 54673-07-3
EINECS: 259-289-9

Bestill nå

Spesifikasjon Testresultater

Appearance Light orange to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR