Cloquintocet mexyl ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI

Synonymer: Cloquintocet-mexyl, 2-Heptyl 2-(5-Chloro-8-quinolinyloxy)acetate, 2-Heptyl 2-[(5-Chloroquinolin-8-yl)oxy]acetate, 2-[(5-Chloroquinolin-8-yl)oxy]acetic Acid 2-Heptyl Ester

C2682-1G
TCIAC2682-1GEA 890 NOK
TCIAC2682-1G
Cloquintocet mexyl ≥98.0% (by HPLC, titration analysis)
Cloquintocet mexyl
Formel: C₁₈H₂₂ClNO₃
MW: 335,83 g/mol
MDL nummer: MFCD01632329
CAS-nummer: 99607-70-2
Merck Indeks: 14,02401

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 68.0 to 72.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR