3-Cyano-1-ethyl-6-hydroxy-4-methyl-2-pyridone ≥98.0% (by HPLC, total nitrogen)

Leverandør: TCI
Danger

Synonymer: 3-Cyano-1-ethyl-6-hydroxy-3-picoline, 1-Ethyl-1,2-dihydro-6-hydroxy-4-methyl-2-oxo-3-pyridinecarbonitrile

C2785-25G
TCIAC2785-25GEA 395 NOK
TCIAC2785-25G
3-Cyano-1-ethyl-6-hydroxy-4-methyl-2-pyridone ≥98.0% (by HPLC, total nitrogen)
3-Cyano-1-ethyl-6-hydroxy-4-methyl-2-pyridone
Formel: C₉H₁₀N₂O₂
MW: 178,19 g/mol
MDL nummer: MFCD00142624
CAS-nummer: 28141-13-1
EINECS: 248-865-5
FN: 3439
ADR: 6.1,III

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow to Light red powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR