Cyclic pifithrin-ɑ-hydrobromide 98,0 HPLC_ASSAY_METHOD

Leverandør: TCI

Synonymer: 2-(p-Tolyl)-5,6,7,8-tetrahydrobenzo[d]imidazo[2,1-b]thiazole hydrobromide, 2-(p-Tolyl)-5,6,7,8-tetrahydroimidazo[2,1-b]benzothiazole Hydrobromide

C2826-20MG C2826-100MG
TCIAC2826-20MGEA 3250 NOK
TCIAC2826-20MG TCIAC2826-100MG
Cyclic pifithrin-ɑ-hydrobromide 98,0 HPLC_ASSAY_METHOD
Cyclic pifithrin-alpha-hydrobromide
Formel: C₁₆H₁₆N₂S
MW: 349.29 g/mol
Lagringstemperatur: Fryseskap
MDL nummer: MFCD02683960
CAS-nummer: 511296-88-1
Merck Indeks: 14,07421

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR