(1S,3R)-(-)-Camphoric acid ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Warning

Synonymer: (1S,3R)-1,2,2-Trimethylcyclopentane-1,3-dicarboxylic acid

C3237-1G
TCIAC3237-1GEA 1350 NOK
TCIAC3237-1G
(1S,3R)-(-)-Camphoric acid ≥98.0% (by GC, titration analysis)
(1S,3R)-(-)-Camphoric acid
Formel: C₁₀H₁₆O₄
MW: 200,23 g/mol
MDL nummer: MFCD00064937
CAS-nummer: 560-09-8

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 186.0 to 190.0 deg-C
Specific rotation [a]20/D -46.0 to -50.0 deg (C=4,EtOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR