3-Cyanopyridine-N-oxide ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Warning

Synonymer: 3-Cyanopyridine 1-oxide, Nicotinonitrile-1-oxide

C3416-1G
TCIAC3416-1GEA 435 NOK
TCIAC3416-1G
3-Cyanopyridine-N-oxide ≥98.0% (by GC, titration analysis)
3-Cyanopyridine-N-oxide
Formel: C₆H₄N₂O
MW: 120,11 g/mol
MDL nummer: MFCD00129037
CAS-nummer: 14906-64-0
EINECS: 238-975-1

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 175.0 to 179.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR