(S,S)-CASA-Na
Leverandør: TCI
Synonymer: Sodium [2,2',2'',2'''-[[(1S,2S)-1,2-Diphenyl-1,2-ethanediyl]bis[[(R)-imino-kappaN]methylidyne]]tetrakis(phenolato-kappaO)]aluminate
C3583-100MG
TCIAC3583-100MGEA 690 NOK
TCIAC3583-100MG
(S,S)-CASA-Na
(S,S)-CASA-Na
Bestill nå
Spesifikasjon Testresultater
Appearance White to Almost white powder to crystal
Suitability for NMR-analysis Effective as chiral shift reagent for 2-Phenylpropionic Acid