(S)-(-)-β-Citronellol ≥98.0% (by GC)

Leverandør: TCI
Warning

C3619-25ML C3619-5ML
TCIAC3619-25MLEA 770 NOK
TCIAC3619-25ML TCIAC3619-5ML
(S)-(-)-β-Citronellol ≥98.0% (by GC)
(S)-(-)-β-Citronellol
Formel: C₁₀H₂₀O
Kokepunkt: 118°C
Flash punkt: 102°C
MDL nummer: MFCD00063214
CAS-nummer: 7540-51-4
EINECS: 231-415-7

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Optical purity (GC) min. 96.0 ee%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR