2'-Deoxycytidine hydrochloride ≥98.0% (by titrimetric analysis)

Leverandør: TCI

D0048-25G D0048-5G
TCIAD0048-25GEA 4225 NOK
TCIAD0048-25G TCIAD0048-5G
2'-Deoxycytidine hydrochloride ≥98.0% (by titrimetric analysis)
2'-Deoxycytidine hydrochloride
Formel: C₉H₁₄ClN₃O₄
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00012840
CAS-nummer: 3992-42-5
EINECS: 223-639-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=20d725fcbb5942779ab7351354b268290355867d&vid=20d725fcbb5942779ab7351354b268290355867d&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (Argentmetric Titration) min. 98.0 %
Specific rotation [a]20/D +55.0 to +59.0 deg (C=1, H2O)
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR