Metylgult

Leverandør: TCI
Danger

Synonymer: Dimethyl yellow, Sudan Yellow, 4-(Dimethylamino)azobenzene, 4-(N,N-Dimethylamino)azobenzene

D0231-25G
TCIAD0231-25GEA 380 NOK
TCIAD0231-25G
Metylgult
Metylgult
Formel: C₁₄H₁₅N₃
MW: 225,29 g/mol
Kokepunkt: 371 °C (1013 hPa)
Smeltepunkt: 113…117 °C
Tetthet: 1,2233 g/cm³ (20 °C)
MDL nummer: MFCD00008308
CAS-nummer: 60-11-7
EINECS: 200-455-7
FN: 2811
ADR: 6.1,III
Merck Indeks: 14,03229
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4a6760e26fb96dc2ace978bf608232882f10ca42&vid=4a6760e26fb96dc2ace978bf608232882f10ca42&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Orange to Brown powder to crystal
Lambda max. 505.0 to 515.0 nm (pH 2.4)
Absorbance min. 0.510 (5 mg/L, pH 2.4, 505.0 to 515.0 nm)
Transition interval (pH) (pale purplish red)2.9 to 4.0 (pale redish yellow)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR