N,N'-Dibenzylethylenediamine diacetate ≥97.0% (by titrimetric analysis)

Leverandør: TCI

Synonymer: N1,N2-Dibenzylethane-1,2-diamine diacetate, Benzathine diacetate, N,N-Dibenzylethylenediamine diacetate

D0280-25G
TCIAD0280-25GEA 285 NOK
TCIAD0280-25G
N,N'-Dibenzylethylenediamine diacetate ≥97.0% (by titrimetric analysis)
N,N'-Dibenzylethylenediamine diacetate
Formel: C₁₆H₂₀N₂
MDL nummer: MFCD00040588
CAS-nummer: 122-75-8
EINECS: 204-572-4

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 116.0 to 120.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR