Dichloramine T ≥96.0% (by titrimetric analysis)

Leverandør: TCI
Danger

Synonymer: N,N-Dichloro-p-toluenesulphonamide, p-Toluenesulphondichloramide

D0318-500G D0318-25G
TCIAD0318-500GEA 6400 NOK
TCIAD0318-500G TCIAD0318-25G
Dichloramine T ≥96.0% (by titrimetric analysis)
Dichloramine T
Formel: C₇H₇Cl₂NO₂S
MDL nummer: MFCD00058929
CAS-nummer: 473-34-7
EINECS: 207-462-4
FN: 1479
ADR: 5.1,I
Merck Indeks: 14,03047

Bestill nå

Spesifikasjon Testresultater

Purity (Iodometric Titration) min. 96.0 %
Melting point 79.0 to 83.0 deg-C

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR