1,3-Dichloro-2-butene cis- and trans-mixture ≥95.0% (by GC)

Leverandør: TCI
Danger

D0351-25G
TCIAD0351-25GEA 4225 NOK
TCIAD0351-25G
1,3-Dichloro-2-butene cis- and trans-mixture ≥95.0% (by GC)
1,3-Dichloro-2-butene
Formel: C₄H₆Cl₂
MW: 125 g/mol
Kokepunkt: 128 °C (1013 hPa)
Smeltepunkt: –98 °C
Tetthet: 1,161 g/cm³ (20 °C)
Flash punkt: 27°C
MDL nummer: MFCD00000861
CAS-nummer: 926-57-8
EINECS: 213-138-3
FN: 1992
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5fa9d6f3cb40963b959f13605aae8a2bb1fd09b1&vid=edc06b8d5f6925fbcdbb90120280f16c179282dc&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 95.0 % (total of isomers)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR