1,6-Diklorheksan ≥98.0% (by GC)

Leverandør: TCI
Warning

D0372-500G D0372-25G
TCIAD0372-500GEA 875 NOK
TCIAD0372-500G TCIAD0372-25G
1,6-Diklorheksan ≥98.0% (by GC)
1,6-Diklorheksan
Formel: ClH₂C(CH₂)₄CH₂Cl
Kokepunkt: 95°C
Smeltepunkt: –13 °C
Tetthet: 1,068 g/cm³ (20 °C)
Flash punkt: 74°C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00001020
CAS-nummer: 2163-00-0
EINECS: 218-491-7
FN: 3082
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4068ac339d137d085040bbe5de524771158cc9aa&vid=965aaf42867d814a7507fd56ba6dae05c36eced6&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR