(±)-Methyl-2,3-dichloropropionate ≥95.0% (by GC)

Leverandør: TCI
Danger

Synonymer: Methyl 2,3-dichloropropanoate

D0408-10G
TCIAD0408-10GEA 325 NOK
TCIAD0408-10G
(±)-Methyl-2,3-dichloropropionate ≥95.0% (by GC)
(±)-Methyl-2,3-dichloropropionate
Formel: C₄H₆Cl₂O₂
MW: 157 g/mol
Kokepunkt: 82°C
Flash punkt: 62°C
MDL nummer: MFCD00000944
CAS-nummer: 3674-09-7
EINECS: 222-940-2
FN: 1760
ADR: 8 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=05655842296452b101c4b2159d822d2ff35e095e&vid=05655842296452b101c4b2159d822d2ff35e095e&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR