Diethylamine hydrochloride ≥98.5% (by titrimetric analysis)

Leverandør: TCI
Warning

D0468-500G D0468-25G
TCIAD0468-500GEA 1550 NOK
TCIAD0468-500G TCIAD0468-25G
Diethylamine hydrochloride ≥98.5% (by titrimetric analysis)
Diethylamine hydrochloride
Formel: C₄H₁₂ClN
Kokepunkt: 320 °C (1013 hPa)
Smeltepunkt: 220…225 °C
Tetthet: 1,0475 g/cm³ (21 °C)
MDL nummer: MFCD00012499
CAS-nummer: 660-68-4
EINECS: 211-541-9
Merck Indeks: 13,03138
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=be70eb5c44e9e91c50b135a3bd500784917127fd&vid=be70eb5c44e9e91c50b135a3bd500784917127fd&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.5 % (calcd.on anh.substance)
Melting point 227.0 to 234.0 deg-C
Solubility in Water almost transparency
Water max. 3.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR