N,N-Diethyl-p-toluidine 98,0 GC, T

Leverandør: TCI
Danger

Synonymer: 4-Diethylaminotoluene, N,N-Diethyl-4-methylaniline

D0533-25ML
TCIAD0533-25MLEA 915 NOK
TCIAD0533-25ML
N,N-Diethyl-p-toluidine 98,0 GC, T
N,N-Diethyl-p-toluidine
Formel: C₁₁H₁₇N
MW: 163.26 g/mol
Kokepunkt: 230°C
Flash punkt: 102°C
MDL nummer: MFCD00015113
CAS-nummer: 613-48-9
EINECS: 210-345-0
FN: 2810
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0ae5ba64106e9c8c0299483d93f0b18bb8c88496&vid=0ae5ba64106e9c8c0299483d93f0b18bb8c88496&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Light orange to Yellow to Green clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR