1,3-Dihydroksynaftalen ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Warning

D0588-1G D0588-5G
TCIAD0588-1GEA 650 NOK
TCIAD0588-1G TCIAD0588-5G
1,3-Dihydroksynaftalen ≥98.0% (by GC, titration analysis)
1,3-Dihydroksynaftalen
Formel: C₁₀H₈O₂
Kokepunkt: 386 °C (1013 hPa)
Smeltepunkt: 124 °C
Tetthet: 1,12 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003965
CAS-nummer: 132-86-5
EINECS: 205-079-7
Merck Indeks: 14,06396
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=85a838ccf838b096bd242fe09d0dc92cfb6250a6&vid=85a838ccf838b096bd242fe09d0dc92cfb6250a6&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 123.0 to 126.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR