N,N'-Diphenyl-p-phenylenediamine ≥95.0% (by titrimetric analysis)

Leverandør: TCI
Warning

D0609-500G D0609-25G D0609-100G
TCIAD0609-500GEA 5525 NOK
TCIAD0609-500G TCIAD0609-25G TCIAD0609-100G
N,N'-Diphenyl-p-phenylenediamine ≥95.0% (by titrimetric analysis)
N,N'-Diphenyl-p-phenylenediamine
Formel: C₁₈H₁₆N₂
Kokepunkt: 250°C
Smeltepunkt: 146…149 °C
Tetthet: 1,21 g/cm³ (20 °C)
MDL nummer: MFCD00003015
CAS-nummer: 74-31-7
EINECS: 200-806-4
Merck Indeks: 14,03329
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e37197a67a0aecf0b4b2454d329042a116595963&vid=e37197a67a0aecf0b4b2454d329042a116595963&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Gray to Brown powder to lump
Purity (GC) min. 90.0 %
Purity (Nonaqueous Titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR