cis-1,2-Dimetylsykloheksan ≥98.0% (by GC)

Leverandør: TCI
Danger

D0696-25ML
TCIAD0696-25MLEA 850 NOK
TCIAD0696-25ML
cis-1,2-Dimetylsykloheksan ≥98.0% (by GC)
cis-1,2-Dimetylsykloheksan
Formel: C₈H₁₆
MW: 112,22 g/mol
Kokepunkt: 130°C
Smeltepunkt: –90 °C
Tetthet: 0,796 g/cm³ (20 °C)
Flash punkt: 12°C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00063621
CAS-nummer: 2207-01-4
EINECS: 218-621-2
FN: 2263
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=79f992d7a9f71be04a7db1f9479ff9d6a8cd296c&vid=79f992d7a9f71be04a7db1f9479ff9d6a8cd296c&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR