N,N-Dimetylformamid ≥99.5% (by GC)

Leverandør: TCI
Danger

D0722-25ML D0722-500ML D0722-100ML
TCIAD0722-25MLEA 230 NOK
TCIAD0722-25ML TCIAD0722-500ML TCIAD0722-100ML
N,N-Dimetylformamid ≥99.5% (by GC)
N,N-Dimetylformamid
Formel: HCON(CH₃)₂
Kokepunkt: 153°C
Smeltepunkt: -61°C
Tetthet: 0,949 g/cm³ (20 °C)
Flash punkt: 60°C (closed cup)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003284
CAS-nummer: 68-12-2
EINECS: 200-679-5
FN: 2265
ADR: 3,III
REACH: 01-2119475605-32
Merck Indeks: 14,03243
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=de9eebe8744933790015429a33188a2f4d7881fa&vid=de9eebe8744933790015429a33188a2f4d7881fa&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless clear liquid
Purity (GC) min. 99.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR