Dimethyl glyoxime disodium salt octahydrate ≥97.0% (W)

Leverandør: TCI
Warning

D0730-25G
TCIAD0730-25GEA 500 NOK
TCIAD0730-25G
Dimethyl glyoxime disodium salt octahydrate ≥97.0% (W)
Dimethyl glyoxime disodium salt octahydrate
Formel: C₄H₆N₂Na₂O₂·8H₂O
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00019957
CAS-nummer: 75006-64-3
EINECS: 262-523-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fee6f91e74a1b1040e1312a963706cc671186c54&vid=fee6f91e74a1b1040e1312a963706cc671186c54&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (Gravimetric Method) min. 97.0 % (calcd.on anh.substance)
Purity (Nonaqueous Titration) min. 90.0 % (calcd.on anh.substance)
Water 44.0 to 49.0 %

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR