N,N-Dimetyl-p-toluidine ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Danger

D0807-25ML D0807-500ML
TCIAD0807-25MLEA 220 NOK
TCIAD0807-25ML TCIAD0807-500ML
N,N-Dimetyl-p-toluidine ≥98.0% (by GC, titration analysis)
N,N-Dimetyl-p-toluidine
Formel: C₉H₁₃N
Kokepunkt: 211°C
Smeltepunkt: –15 °C
Tetthet: 0,9 g/cm³ (20 °C)
Flash punkt: 90°C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00008316
CAS-nummer: 99-97-8
EINECS: 202-805-4
FN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=33fc9198bcc48cc0b676c199a84491567db6d8e0&vid=33fc9198bcc48cc0b676c199a84491567db6d8e0&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR