4,4'-Dinitrobiphenyl ≥99.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 4,4'-Dinitro-1,1'-biphenyl

D0826-5G
TCIAD0826-5GEA 2025 NOK
TCIAD0826-5G
4,4'-Dinitrobiphenyl ≥99.0% (by GC)
4,4'-Dinitrobiphenyl
Formel: C₁₂H₈N₂O₄
MW: 244.21 g/mol
MDL nummer: MFCD00039744
CAS-nummer: 1528-74-1
EINECS: 216-210-2

Bestill nå

Spesifikasjon Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 99.0 %
Melting point 240.0 to 244.0 deg-C
Solubility in hot Toluene almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR