Diphenyl phosphite ≥75.0% (by HPLC) (contains ca. 5% Phenol)

Leverandør: TCI
Danger

Synonymer: Diphenoxyphosphine oxide, NSC 43786, Diphenyl phosphonate

D0907-25G D0907-500G D0907-100G
TCIAD0907-25GEA 245 NOK
TCIAD0907-25G TCIAD0907-500G TCIAD0907-100G
Diphenyl phosphite ≥75.0% (by HPLC) (contains ca. 5% Phenol)
Diphenyl phosphite
Formel: C₁₂H₁₁O₃P
Kokepunkt: 148°C
Smeltepunkt: 12°C
Flash punkt: 186°C
MDL nummer: MFCD00044497
CAS-nummer: 4712-55-4
EINECS: 225-202-8
FN: 1760
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0f48535dff456396074a2370dd67837203e6614a&vid=0f48535dff456396074a2370dd67837203e6614a&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (HPLC) min. 75.0 area%
Phenol 3.0 and le 6.0 w/w%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR