2,5-Diphenyl-3-(4-styrylphenyl)tetrazolium Chloride ≥96.0% (by titrimetric analysis)

Leverandør: TCI

D0915-1G
TCIAD0915-1GEA 1350 NOK
TCIAD0915-1G
2,5-Diphenyl-3-(4-styrylphenyl)tetrazolium Chloride ≥96.0% (by titrimetric analysis)
2,5-Diphenyl-3-(4-styrylphenyl)tetrazolium chloride
Formel: C₂₇H₂₁ClN₄
MDL nummer: MFCD00050315
CAS-nummer: 24387-36-8
EINECS: 246-215-5

Bestill nå

Spesifikasjon Testresultater

Appearance Light orange to Yellow to Green powder to crystal
Purity (Nonaqueous Titration) min. 96.0 %
Melting point (Decomposition) 219.0 to 227.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR