β-Nicotinamide adenine dinucleotide, oxidised form (NAD, oxidised form) hydrate ≥95.0% (by HPLC) for biochemical research

Leverandør: TCI

Synonymer: Cozymase, beta-DPN, Coenzyme I, beta-Nicotinamide Adenine dinucleotide hydrate, oxidized form, beta-Diphosphopyridine Nucleotide, beta-NAD

D0919-1G D0919-5G
TCIAD0919-1GEA 770 NOK
TCIAD0919-1G TCIAD0919-5G
β-Nicotinamide adenine dinucleotide, oxidised form (NAD, oxidised form) hydrate ≥95.0% (by HPLC) for biochemical research
β-Nicotinamide adenine dinucleotide, oxidised form (NAD, oxidised form) hydrate
Formel: C₂₁H₂₇N₇O₁₄P₂·H₂O
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00036253
CAS-nummer: 53-84-9
EINECS: 200-184-4
Merck Indeks: 14,06344

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 95.0 area%
abs-ratio (a/b) 0.80 to 0.86 (E250/E260)
abs-ratio (2a/2b) 0.19 to 0.23 (E280/E260)
Molar extinction coefficient 17,000 to 18,500 (H2O, 260.0 nm) (calcd.on anh.substance)

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR