Natriumdodecylsulfat ≥85.0% (by titrimetric analysis)

Leverandør: TCI
Warning

Synonymer: Dodecyl sulphate sodium salt, Sodium dodecyl sulfate, Dodecyl sulphuric acid sodium salt, Lauryl sulphate sodium salt, Sodium lauryl sulphate

D0996-25G D0996-500G
TCIAD0996-25GEA 220 NOK
TCIAD0996-25G TCIAD0996-500G
Natriumdodecylsulfat ≥85.0% (by titrimetric analysis)
Natriumdodecylsulfat
Formel: CH₃(CH₂)₁₁OSO₃Na
Smeltepunkt: 207°C
Tetthet: 1,1 g/cm³ (20 °C)
Flash punkt: ≥ 100 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00036175
CAS-nummer: 151-21-3
EINECS: 205-788-1
ADR: 4.1,II
Merck Indeks: 14,08636
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3af268a7380d51ba135b686612a8042cf953b348&vid=6ad5f4c94cb0bfebeeb0ae8c15566e9272a07b2d&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 50.0 %
Purity (Neutralization back titration) min. 85.0 %
Average molecular weight 285 to 310

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR