Diphenyl chlorophosphate ≥95.0% (by GC)

Leverandør: TCI
Danger

D1059-25G D1059-500G D1059-100G
TCIAD1059-25GEA 245 NOK
TCIAD1059-25G TCIAD1059-500G TCIAD1059-100G
Diphenyl chlorophosphate ≥95.0% (by GC)
Diphenyl chlorophosphate
Formel: C₁₂H₁₀ClO₃P
Kokepunkt: 316°C
MDL nummer: MFCD00003030
CAS-nummer: 2524-64-3
EINECS: 219-759-6
FN: 3265
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5abe6e49bf56eac9643a513587319cc394791431&vid=5abe6e49bf56eac9643a513587319cc394791431&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR