9-(4'-Dimethylaminophenyl)-2,6,7-trihydroxyfluorone sulphate hydrate ≥96.0% (by HPLC)

Leverandør: TCI

Synonymer: Dimethylaminophenylfluorone (so called)

D1118-1G
TCIAD1118-1GEA 1300 NOK
TCIAD1118-1G
9-(4'-Dimethylaminophenyl)-2,6,7-trihydroxyfluorone sulphate hydrate ≥96.0% (by HPLC)
9-(4'-Dimethylaminophenyl)-2,6,7-trihydroxyfluorone sulphate hydrate
Formel: C₂₁H₁₇NO₅
MDL nummer: MFCD00035151
CAS-nummer: 71501-19-4
EINECS: 275-549-4

Bestill nå

Spesifikasjon Testresultater

Appearance Orange to Brown powder to crystal
Purity (HPLC) min. 96.0 area%
Water max. 15.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR