2,5-Diphenyl-p-benzoquinone ≥99.0% (by GC)

Leverandør: TCI

D1166-25G
TCIAD1166-25GEA 4850 NOK
TCIAD1166-25G
2,5-Diphenyl-p-benzoquinone ≥99.0% (by GC)
2,5-Diphenyl-p-benzoquinone
Formel: C₁₈H₁₂O₂
MDL nummer: MFCD00001600
CAS-nummer: 844-51-9
EINECS: 212-680-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=effed1304c12bac1a30a4594ffb9060708e9bfe2&vid=effed1304c12bac1a30a4594ffb9060708e9bfe2&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 99.0 %
Melting point 216.0 to 220.0 deg-C
Solubility in hot Toluene almost transparency

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR