3,4-Dimetylheksan ≥97.0% (by GC)

Leverandør: TCI
Danger

D1228-5ML
TCIAD1228-5MLEA 1925 NOK
TCIAD1228-5ML
3,4-Dimetylheksan ≥97.0% (by GC)
3,4-Dimetylheksan
Formel: C₈H₁₈
MW: 114,23 g/mol
Kokepunkt: 117°C
Smeltepunkt: –64 °C
Tetthet: 0,7152 g/cm³ (20 °C)
Flash punkt: 13°C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00039927
CAS-nummer: 583-48-2
EINECS: 209-504-7
FN: 1262
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d9be0a3efdb46980d0deee4779f7e8b3e213fe56&vid=d9be0a3efdb46980d0deee4779f7e8b3e213fe56&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR