Sodium-2,4-dichlorophenoxyacetate monohydrate ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Warning

Synonymer: Na-2,4-D, 2,4-Dichlorophenoxyacetic Acid Sodium Salt

D1319-25G D1319-500G
TCIAD1319-25GEA 220 NOK
TCIAD1319-25G TCIAD1319-500G
Sodium-2,4-dichlorophenoxyacetate monohydrate ≥98.0% (by HPLC, titration analysis)
Sodium-2,4-dichlorophenoxyacetate monohydrate
Formel: C₈H₆Cl₂O₃
MW: 243.01 g/mol
MDL nummer: MFCD00068284
CAS-nummer: 2702-72-9
EINECS: 220-290-4

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 % (calcd.on anh.substance)
Water 6.0 to 8.0 %

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR