2,2-Dichloropropionic acid ≥95.0% (by GC)

Leverandør: TCI
Danger

D1334-1G
TCIAD1334-1GEA 420 NOK
TCIAD1334-1G
2,2-Dichloropropionic acid ≥95.0% (by GC)
2,2-Dichloropropionic acid
Formel: C₃H₄Cl₂O₂
MW: 142.97 g/mol
MDL nummer: MFCD00004178
CAS-nummer: 75-99-0
EINECS: 200-923-0
Merck Indeks: 14,02802

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR