Chloro(methyl)(diphenyl)silane ≥95.0% (by GC)

Leverandør: TCI
Danger

D1390-25ML
TCIAD1390-25MLEA 1025 NOK
TCIAD1390-25ML
Chloro(methyl)(diphenyl)silane ≥95.0% (by GC)
Chloro(methyl)(diphenyl)silane
Formel: C₁₃H₁₃ClSi
Flash punkt: 141°C
MDL nummer: MFCD00000498
CAS-nummer: 144-79-6
EINECS: 205-639-0
FN: 2987
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c57431d2bd884ac6df712f0fe006924bc98d0f12&vid=c57431d2bd884ac6df712f0fe006924bc98d0f12&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR