(Dimethylamino)acetaldehyde diethylacetal ≥97.0% (by GC, titration analysis)

Leverandør: TCI
Warning

Synonymer: 2,2-Diethoxy-N,N-dimethylethanamine

D1703-25ML
TCIAD1703-25MLEA 930 NOK
TCIAD1703-25ML
(Dimethylamino)acetaldehyde diethylacetal ≥97.0% (by GC, titration analysis)
(Dimethylamino)acetaldehyde diethylacetal
Formel: C₈H₁₉NO₂
MW: 161.25 g/mol
Kokepunkt: 60°C
Flash punkt: 45°C
MDL nummer: MFCD00009232
CAS-nummer: 3616-56-6
EINECS: 222-800-0
FN: 1993
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=43a70c01cca6e1effa4fad57def58e4327c65025&vid=43a70c01cca6e1effa4fad57def58e4327c65025&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR