(Dimethylamino)acetaldehyde dimethylacetal ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Warning

Synonymer: 2,2-Dimethoxy-N,N-dimethylethanamine, N-(2,2-Dimethoxyethyl)dimethylamine

D1704-25ML
TCIAD1704-25MLEA 985 NOK
TCIAD1704-25ML
(Dimethylamino)acetaldehyde dimethylacetal ≥98.0% (by GC, titration analysis)
(Dimethylamino)acetaldehyde dimethylacetal
Formel: C₆H₁₅NO₂
MW: 133.1888 g/mol
Kokepunkt: 81°C
Flash punkt: 32°C
MDL nummer: MFCD00010205
CAS-nummer: 38711-20-5
EINECS: 254-097-1
FN: 1993
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=28549fb98eac96c387234898ad9c2782b757e3ae&vid=28549fb98eac96c387234898ad9c2782b757e3ae&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR