2,2'-Dimethylbiphenyl ≥95.0% (by GC)

Leverandør: TCI

Synonymer: 2,2'-Bitolyl, 2,2'-Dimethyl-1,1'-biphenyl

D1717-1G D1717-5G
TCIAD1717-1GEA 460 NOK
TCIAD1717-1G TCIAD1717-5G
2,2'-Dimethylbiphenyl ≥95.0% (by GC)
2,2'-Dimethylbiphenyl
Formel: C₁₄H₁₄
MW: 182.27 g/mol
Kokepunkt: 259°C
Smeltepunkt: 18
MDL nummer: MFCD00048075
CAS-nummer: 605-39-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=53191249a92547b0a84bfb091b64bae6720ac8b0&vid=53191249a92547b0a84bfb091b64bae6720ac8b0&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR