1-Chloro-3,4-dinitrobenzene ≥90.0% (by GC)

Leverandør: TCI
Danger

D1900-25G
TCIAD1900-25GEA 1050 NOK
TCIAD1900-25G
1-Chloro-3,4-dinitrobenzene ≥90.0% (by GC)
1-Chloro-3,4-dinitrobenzene
Formel: C₆H₃ClN₂O₄
MW: 202,55 g/mol
Kokepunkt: 315°C
Smeltepunkt: 40…41 °C
Tetthet: 1,687 g/cm³ (20 °C)
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00007212
CAS-nummer: 610-40-2
EINECS: 210-223-7
FN: 3441
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=78664e04d626e6ba1ad3827427fd77593ace29d5&vid=78664e04d626e6ba1ad3827427fd77593ace29d5&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Brown to Dark green powder to lump
Purity (GC) min. 90.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR